Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Acetyl-2-methylbenzoic acid
4-Acetyl-2-methylbenzoic acid
4-Acetyl-2-methylbenzoic acid
4-Acetyl-2-methylbenzoic acid
APOSOR401070-1G
:
Apollo Scientific
:
4-Acetyl-2-methylbenzoic acid
APOSOR401070-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
178.185 g/mol
:
Ambient
:
55860-35-0
:
C₁₀H₁₀O₃
:
MFCD20639525