
Methyl 3-bromo-2-(bromomethyl)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:337536-14-8
:
Formula:C₉H₈Br₂O₂
MDL Number:MFCD06804569
:307.967 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together