Order Entry
Ireland
ContactUsLinkComponent
4-(2-(Hydroxymethyl)phenyl)-1-methyl-3-phenylpiperidin-4-ol ≥98%
4-(2-(Hydroxymethyl)phenyl)-1-methyl-3-phenylpiperidin-4-ol ≥98%
Catalog # APOSOR370047-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-(2-(Hydroxymethyl)phenyl)-1-methyl-3-phenylpiperidin-4-ol ≥98%
Catalog # APOSOR370047-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :297.391 g/mol
Storage Temperature:Ambient
CAS Number:218288-32-5
Formula:C₁₉H₂₃NO₂
MDL Number:MFCD18207051