
2-Chloro-5-methoxy-3-nitropyridine
Catalog # APOSOR345499-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:188.569 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1003711-55-4
Formula:C₆H₅ClN₂O₃
MDL Number:MFCD09835251