Methyl 5-methoxy-3-methyl-1H-indole-2-carboxylate, tech
Catalog # APOSOR33647-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.237 g/mol
Storage Temperature:Ambient
CAS Number:480996-91-6
Formula:C₁₂H₁₃NO₃
MDL Number:MFCD06496116