Order Entry
Ireland
ContactUsLinkComponent
2-Oxo-6-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
2-Oxo-6-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR33353-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Oxo-6-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR33353-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :234.253 g/mol
Storage Temperature:Ambient
CAS Number:339109-43-2
Formula:C₁₅H₁₀N₂O
MDL Number:MFCD00202806