Order Entry
Ireland
ContactUsLinkComponent
2-[3-(2,3-Dihydro-1H-inden-5-yl)-1H-pyrazol-1-yl]ethan-1-ol, tech
2-[3-(2,3-Dihydro-1H-inden-5-yl)-1H-pyrazol-1-yl]ethan-1-ol, tech
Catalog # APOSOR33164-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[3-(2,3-Dihydro-1H-inden-5-yl)-1H-pyrazol-1-yl]ethan-1-ol, tech
Catalog # APOSOR33164-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.29 g/mol
Storage Temperature:Ambient
CAS Number:956754-78-2
Formula:C₁₄H₁₆N₂O
MDL Number:MFCD03848645