6-(2,3-Dihydro-1H-inden-5-yl)-2,3,4,5-tetrahydropyridazin-3-one, tech
Catalog # APOSOR33036-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:214.263 g/mol
Storage Temperature:Ambient
CAS Number:866050-09-1
Formula:C₁₃H₁₄N₂O
MDL Number:MFCD03787574