N-Phenyl-2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetamide, tech
Catalog # APOSOR32848-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.338 g/mol
Storage Temperature:Ambient
CAS Number:338761-98-1
Formula:C₁₇H₁₈N₂O
MDL Number:MFCD00215028