2-[(Furan-2-yl)methylidene]-2,3-dihydro-1H-indene-1,3-dione, tech
Catalog # APOSOR32469-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.212 g/mol
Storage Temperature:Ambient
CAS Number:29874-41-7
Formula:C₁₄H₈O₃
MDL Number:MFCD00219001