Order Entry
Ireland
ContactUsLinkComponent
Methyl 1-oxo-2-phenyl-1,2-dihydroisoquinoline-4-carboxylate, tech
Methyl 1-oxo-2-phenyl-1,2-dihydroisoquinoline-4-carboxylate, tech
Catalog # APOSOR32173-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 1-oxo-2-phenyl-1,2-dihydroisoquinoline-4-carboxylate, tech
Catalog # APOSOR32173-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.29 g/mol
Storage Temperature:Ambient
CAS Number:73349-64-1
Formula:C₁₇H₁₃NO₃
MDL Number:MFCD00140322