Order Entry
Ireland
ContactUsLinkComponent
3,4,4,6-Tetramethyl-1,2,3,4-tetrahydropyrimidine-2-thione, tech
3,4,4,6-Tetramethyl-1,2,3,4-tetrahydropyrimidine-2-thione, tech
Catalog # APOSOR32032-1G
Supplier:  Apollo Scientific
CAS Number:  
3,4,4,6-Tetramethyl-1,2,3,4-tetrahydropyrimidine-2-thione, tech
Catalog # APOSOR32032-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.275 g/mol
Storage Temperature:Ambient
CAS Number:58544-48-2
Formula:C₈H₁₄N₂S
MDL Number:MFCD00463296