Order Entry
Ireland
ContactUsLinkComponent
Methyl 3-formyl-1-methyl-1h-pyrazole-5-carboxylate 97%
Methyl 3-formyl-1-methyl-1h-pyrazole-5-carboxylate 97%
Catalog # APOSOR314008-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-formyl-1-methyl-1h-pyrazole-5-carboxylate 97%
Catalog # APOSOR314008-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :168.15 g/mol
Storage Temperature:Ambient
CAS Number:1031351-95-7
Formula:C₇H₈N₂O₃
MDL Number:MFCD22376784