Order Entry
Ireland
ContactUsLinkComponent
 
Ethyl 6-(chloromethyl)-4-(2-chlorophenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Catalog # APOSOR310998-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 6-(chloromethyl)-4-(2-chlorophenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Catalog # APOSOR310998-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :329.179 g/mol
Storage Temperature:Ambient
CAS Number:325479-50-3
Formula:C₁₄H₁₄Cl₂N₂O₃
MDL Number:MFCD00749774