Methyl 6-bromo-1-(propan-2-yl)-1H-indazole-4-carboxylate
Catalog # APOSOR310529-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:297.148 g/mol
Storage Temperature:Ambient
CAS Number:1346702-52-0
Formula:C₁₂H₁₃BrN₂O₂
MDL Number:MFCD26407262