2-Methylpropane-1,2-diol
Catalog # APOSOR310412-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:90.121 g/mol
Boiling Pt:177…178 °C
Density:0,997 g/cm³ (25 °C)
EINECS:209-198-5
:GHS07
Storage Temperature:Ambient
CAS Number:558-43-0
Formula:C₄H₁₀O₂
MDL Number:MFCD00971926