
[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:299430-87-8
:
Formula:C₁₉H₂₁NO₄
MDL Number:MFCD04974286
:327.374 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together