Order Entry
Ireland
ContactUsLinkComponent
(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-2-phenyl-ethanol
(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-2-phenyl-ethanol
Catalog # APOSOR307634-1G
Supplier:  Apollo Scientific
CAS Number:  
(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-2-phenyl-ethanol
Catalog # APOSOR307634-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :313.391 g/mol
Storage Temperature:Ambient
CAS Number:548443-18-1
Formula:C₁₉H₂₃NO₃
MDL Number:MFCD10703291