
(cis-1-Amino-indan-2-yl)methanol
Catalog # APOSOR307619-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:163.216 g/mol
Boiling Pt:108…110 °C (1 mm Hg)
Melting Pt:81…83 °C
:GHS07
Storage Temperature:Ambient
CAS Number:55270-04-7
Formula:C₁₀H₁₃NO
MDL Number:MFCD07783060