
1-tert-Butyl 2-methyl (2R,4R)-4-aminopyrrolidine-1,2-dicarboxylate 95%
Catalog # APOSOR30678-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:244.288 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1146160-08-8
Formula:C₁₁H₂₀N₂O₄
MDL Number:MFCD08704544