3-Oxoindane-5-carboxylic acid 97%
Catalog # APOSOR30552-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:176.169 g/mol
Melting Pt:256…260 °C
Storage Temperature:Ambient
CAS Number:60031-08-5
Formula:C₁₀H₈O₃
MDL Number:MFCD02179295