Order Entry
Ireland
ContactUsLinkComponent
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indoline
Irritant
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indoline
Catalog # APOSOR305264-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indoline
Catalog # APOSOR305264-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :245.13 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:2304634-69-1
 :1253912-15-0
Formula:C₁₄H₂₀BNO₂
MDL Number:MFCD16995802