Order Entry
Ireland
ContactUsLinkComponent
(1R)-5-tert-Butyl-2,3-dihydro-1H-inden-1-amine ≥ ee 99%
(1R)-5-tert-Butyl-2,3-dihydro-1H-inden-1-amine ≥ ee 99%
Catalog # APOSOR303308-500MG
Supplier:  Apollo Scientific
CAS Number:  
(1R)-5-tert-Butyl-2,3-dihydro-1H-inden-1-amine ≥ ee 99%
Catalog # APOSOR303308-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :189.297 g/mol
Storage Temperature:Ambient
CAS Number:808756-83-4
Formula:C₁₃H₁₉N
MDL Number:MFCD11515603