Order Entry
Ireland
ContactUsLinkComponent
6-Hydroxyindole, N-BOC protected 95%
6-Hydroxyindole, N-BOC protected 95%
Catalog # APOSOR2572-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Hydroxyindole, N-BOC protected 95%
Catalog # APOSOR2572-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :233.263 g/mol
Storage Temperature:Ambient
CAS Number:898746-82-2
Formula:C₁₃H₁₅NO₃
MDL Number:MFCD08272252