Order Entry
Ireland
ContactUsLinkComponent
 
 
Diethyl 2-aminomalonate, N-BOC protected 97%
  APOSOR22746-5G
 :  
 
Diethyl 2-aminomalonate, N-BOC protected 97%
  APOSOR22746-5G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

 :
 :275.298 g/mol
 :218 °C
 :1,079 g/cm³ (25 °C)
 :113 °C
 :GHS07
 :Ambient
 :102831-44-7
 :C₁₂H₂₁NO₆
 :MFCD00239423