
4-(1-Methyl-1H-indol-3-yl)but-3-en-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:169057-08-3
:
Formula:C₁₃H₁₃NO
MDL Number:MFCD00218450
:199.248 g/mol
Melting Pt:104…106 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together