
Methyl 5-chloro-4-(2-methoxy-2-oxoethyl)-2-methyl-1H-pyrrole-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:77978-86-0
:
Formula:C₁₀H₁₂ClNO₄
MDL Number:MFCD00179171
:245.66 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together