Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N1-(8-quinolyl)-2-chloroacetamide
N1-(8-quinolyl)-2-chloroacetamide
N1-(8-quinolyl)-2-chloroacetamide
N1-(8-quinolyl)-2-chloroacetamide
APOSOR21196-1G
:
Apollo Scientific
:
N1-(8-quinolyl)-2-chloroacetamide
APOSOR21196-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
:
220.655 g/mol
:
Ambient
:
32889-11-5
:
C₁₁H₉ClN₂O
:
MFCD00218186