2,4,6-Tribromobenzene-1,3-diamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:62477-06-9
:
Formula:C₆H₅Br₃N₂
MDL Number:MFCD00466367
:344.829 g/mol
Melting Pt:158…162 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together