
6-Benzyloxy-3-iodo-1H-indole, N-BOC protected 95%
Catalog # APOSOR1724-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:449.282 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:914349-30-7
Formula:C₂₀H₂₀INO₃
MDL Number:MFCD05864773