5-(Benzyloxy)-3-iodo-1H-indole, N-BOC protected 95+%
Catalog # APOSOR1722-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:449.282 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:914349-29-4
Formula:C₂₀H₂₀INO₃
MDL Number:MFCD05864771