
5-(Benzyloxy)-3-bromo-1H-indole, N-BOC protected 95%
Catalog # APOSOR1721-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:402.282 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:914349-28-3
Formula:C₂₀H₂₀BrNO₃
MDL Number:MFCD05864770