Order Entry
Ireland
ContactUsLinkComponent
4-Bromoindan-2-one 97%
Irritant
4-Bromoindan-2-one 97%
Catalog # APOSOR17022-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromoindan-2-one 97%
Catalog # APOSOR17022-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :211.055 g/mol
Melting Pt:74…75 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:846032-36-8
Formula:C₉H₇BrO
MDL Number:MFCD09744095