Order Entry
Ireland
ContactUsLinkComponent
1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)imidazolidin-2-one
1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)imidazolidin-2-one
Catalog # APOSOR16160-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)imidazolidin-2-one
Catalog # APOSOR16160-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :317.383 g/mol
Storage Temperature:Ambient
CAS Number:1209101-05-2
Formula:C₁₇H₂₃N₃O₃
MDL Number:MFCD11109514