Order Entry
Ireland
ContactUsLinkComponent
1-(2-Chloroethyl)-3-(7-nitro-1,1,3,3,6-pentamethylindan-5-yl)urea
1-(2-Chloroethyl)-3-(7-nitro-1,1,3,3,6-pentamethylindan-5-yl)urea
Catalog # APOSOR16158-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Chloroethyl)-3-(7-nitro-1,1,3,3,6-pentamethylindan-5-yl)urea
Catalog # APOSOR16158-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :353.844 g/mol
Storage Temperature:Ambient
CAS Number:1211016-69-1
Formula:C₁₇H₂₄ClN₃O₃
MDL Number:MFCD11109512