
3-Formyl-6-(methoxycarbonyl)-1H-indole, N-BOC protected
Catalog # APOSOR1457-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:303.31 g/mol
Melting Pt:166…168 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:850374-95-7
Formula:C₁₆H₁₇NO₅
MDL Number:MFCD06797500