5-(2-Aminothiazol-5-yl)sulphanyl-2-methoxy-4-methylbenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:439578-97-9
:
Formula:C₁₂H₁₂N₂O₃S₂
MDL Number:MFCD19443226
:296.365 g/mol
Storage Temperature:Ambient
Frequently Bought Together