Ethyl [(3-formyl-1-methyl-1H-indol-2-yl)thio]acetate
Catalog # APOSOR111483-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:277.339 g/mol
Storage Temperature:Ambient
CAS Number:2197055-31-3
Formula:C₁₄H₁₅NO₃S
MDL Number:MFCD31380614