
Fmoc-NH-PEG8-CH2COOH
Catalog # APOSBIPG1413-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:649.726 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:868594-52-9
Formula:C₃₃H₄₇NO₁₂
MDL Number:MFCD27635163