
Fmoc-N-amido-PEG24-acid
Catalog # APOSBIPG1407-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:1368.593 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:2170484-59-8
Formula:C₆₆H₁₁₃NO₂₈
MDL Number:MFCD11041159