
Azido-PEG10-acid
Catalog # APOSBIPG1111-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:555.616 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1644163-57-4
Formula:C₂₃H₄₅N₃O₁₂
MDL Number:MFCD26127811