0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
4-Azidophenyl glyoxal hydrate
4-Azidophenyl glyoxal hydrate
4-Azidophenyl glyoxal hydrate
APOSBIPA108-100MG
:
Apollo Scientific
:
4-Azidophenyl glyoxal hydrate
APOSBIPA108-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
193.159 g/mol
:
95…122 °C
:
GHS07
:
Refrigerator
:
1196151-49-1
:
C₈H₅N₃O₂·H₂O
:
MFCD00150595