Order Entry
Ireland
ContactUsLinkComponent
PMPI (4-Maleimidophenyl isocyanate)
Irritant
Health
PMPI (4-Maleimidophenyl isocyanate)
Catalog # APOSBICL223-50MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Health
PMPI (4-Maleimidophenyl isocyanate)
Catalog # APOSBICL223-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :214.177 g/mol
Melting Pt:120…125 °C
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:123457-83-0
Formula:C₁₁H₆N₂O₃
MDL Number:MFCD00270900