Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Catalog # APOSBICL205-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:482.205 g/mol
:GHS07
Storage Temperature:Freezer
:MFCD00054977
Formula:C₁₃H₁₁IN₂O₈S
MDL Number:MFCD01861953