5-Chloro-DL-tryptophan
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:154-07-4
:
Formula:C₁₁H₁₁ClN₂O₂
MDL Number:MFCD01074499
Melting Pt:278…279 °C
:238.6700 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together