
Terpineol, mixture of isomers ≥80.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:8000-41-7
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00166983
:154.25 g/mol
:GHS07
Specification Test Results
- Assay (GC, area%) (isomers):min 80,0%
- Identification (IR):passes test
- Density (20/4):0,932 - 0,936
Specification Test Results
Related Information
Frequently Bought Together