BIX2188 Z-Isomer 95%
ACRO467971000
:
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
- Appearance:White to yellow Powder or solid
- HPLC:≥94%
- MS:complies
- NMR (1H-NMR):Consistent with structure
: BIX-02188, BIX02188, (Z)-3-(((3-((dimethylamino)methyl)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxamide
:
:412.493 g/mol
:GHS07
:Freezer
:334949-59-6
:1094614-84-2
:C₂₅H₂₄N₄O₂
:MFCD18157583