Hydrochloric acid 0,05 mol/l (0,05 N) (20°C), Titripur®, Supelco®
Catalog # 1.60327.1003
Supplier: Merck
CAS Number:
Specifications
- Pack type:Bottle with RFID tag
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:36.46 g/mol
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324