Order Entry
Ireland
ContactUsLinkComponent
Impregnated biocide wipes, low residue quaternary ammonium compound with WFI, Klerwipe™
Impregnated biocide wipes, low residue quaternary ammonium compound with WFI, Klerwipe™
Catalog # SHIE6600052
Impregnated biocide wipes, low residue quaternary ammonium compound with WFI, Klerwipe™
Catalog # SHIE6600052
Specifications
  • Packed:
    Resealable pouch, 20×10 wipes
  • Description:
    Sterile low residue quaternary compounds wipes
  • L×W:
    200×200 mm
  • Pk:
    200
Specifications
About this item
Sterile heavy weight, pre-laundered, ultrasonically sealed edge 100% polyester wipes impregnated with a very low residue quaternary ammonium compound.
  • Low residue accumulation effect
  • Ready-to-use
  • Resealable pouch
  • Has Klertear on outer wraps
  • For small surfaces
The biocide blend is 0,2 µm filtered and sterile filled under Grade A laminar flow and double bagged in a Grade B cleanroom.
Certifications: Bactericide: EN 1276, EN 13697, EN 16615; Yeasticide: EN 1650, EN 13697, EN 16615; Virucidal: RKI/DVV.
Caution: Use biocides safely. Always read the label and product information before use.
*The product(s) on this page are outside the scope of the EU Directive 2019/904