
Ethyl viologen diperchlorate, Sigma-Aldrich®
Catalog # 384100-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:413.21 g/mol
Melting Pt:270 °C (dec., lit.)
:GHS07
Storage Temperature:Ambient
CAS Number:36305-51-8
Formula:C₁₄H₁₈Cl₂N₂O₈
MDL Number:MFCD00134591